accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONSBr

CAS RN 194851-19-9

IUPAC Name: 4-[(4-bromothiophen-2-yl)methyl]morpholine
Synonyms: 4-[(4-bromothiophen-2-yl)methyl]morpholine
Molecular Weight (g/mol): 262.17
Molecular Formula: C9H12BrNOS
InChi Key: XYQVGDMXJNORKK-UHFFFAOYSA-N
SMILES: BrC1=CSC(CN2CCOCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results