accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1949-45-7

IUPAC Name: 3-acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzoic acid
Synonyms: metrizoic acid acidum metrizoicum acide metrizoique 甲泛影酸
Molecular Weight (g/mol): 627.94
Molecular Formula: C12H11I3N2O4
InChi Key: GGGDNPWHMNJRFN-UHFFFAOYSA-N
SMILES: CN(C(C)=O)C1=C(I)C(NC(C)=O)=C(I)C(C(O)=O)=C1I
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results