accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19499-60-6

CH3HNNOOHCl

CAS RN 19499-60-6

IUPAC Name: methyl[(4-nitrophenyl)methyl]amine hydrochloride
Synonyms: methyl[(4-nitrophenyl)methyl]amine hydrochloride
Molecular Weight (g/mol): 202.64
Molecular Formula: C8H11ClN2O2
InChi Key: LADGAFKIYVCZGN-UHFFFAOYSA-N
SMILES: Cl.CNCC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results