accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 195503-40-3

IUPAC Name: thiane-4-carbonitrile
Synonyms: thiane-4-carbonitrile
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: DQKVZWAEWJUKAX-UHFFFAOYSA-N
SMILES: N#CC1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results