CAS RN 1956-11-2
IUPAC Name:
4-nitrophenyl dodecanoate
Synonyms:
4-nitrophenyl dodecanoate
Molecular Weight (g/mol):
321.42
Molecular Formula:
C18H27NO4
InChi Key:
YNGNVZFHHJEZKD-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O