accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 195604-39-8

IUPAC Name: 1-cyclopropylethan-1-amine
Synonyms: 1-cyclopropylethanamine
Molecular Weight (g/mol): 85.15
Molecular Formula: C5H11N
InChi Key: IXCXVGWKYIDNOS-UHFFFAOYNA-N
SMILES: CC(N)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results