accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSBr

CAS RN 19614-16-5

IUPAC Name: 1-bromo-2-(methylsulfanyl)benzene
Synonyms: 1-bromo-2-(methylsulfanyl)benzene
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H7BrS
InChi Key: ALAQDUSTXPEHMH-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1Br
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)