accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1967-25-5

IUPAC Name: (4-bromophenyl)urea
Synonyms: P-bromophenylurea
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: PFQUUCXMPUNRLA-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=C(Br)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results