accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19694-10-1

IUPAC Name: 3-amino-4-chlorobenzamide
Synonyms: 3-amino-4-chlorobenzamide
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: QHMDKGRWJVOUFU-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(Cl)C(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results