accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(R)(S)OHOHNOHOO

CAS RN 19728-63-3

IUPAC Name: (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoate
Synonyms: (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoate
Molecular Weight (g/mol): 252.25
Molecular Formula: C12H14NO5
InChi Key: IPJUIRDNBFZGQN-SCZZXKLOSA-M
SMILES: C[C@@H](O)[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results