accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 1973-22-4

IUPAC Name: 1-bromo-2-ethylbenzene
Synonyms: 1-bromo-2-ethylbenzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: HVRUGFJYCAFAAN-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1Br
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results