accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONNO

CAS RN 197638-83-8

IUPAC Name: tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(4-formylphenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 290.36
Molecular Formula: C16H22N2O3
InChi Key: KHORERZDMJTBMR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results