accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19764-30-8

IUPAC Name: 2-acetamido-4-methylpentanoic acid
Synonyms: acetylleucine
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: WXNXCEHXYPACJF-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(C)=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)