accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOH3COCl

CAS RN 197719-27-0

IUPAC Name: dimethyl-1,3-oxazole-4-carbonyl chloride
Synonyms: dimethyl-1,3-oxazole-4-carbonyl chloride
Molecular Weight (g/mol): 159.57
Molecular Formula: C6H6ClNO2
InChi Key: XZXCVPKMCPXVFF-UHFFFAOYSA-N
SMILES: CC1=NC(C(Cl)=O)=C(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results