accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19785-39-8

IUPAC Name: 2-(4-methyl-1,3-thiazol-2-yl)acetonitrile
Synonyms: 2-(4-methyl-1,3-thiazol-2-yl)acetonitrile
Molecular Weight (g/mol): 138.19
Molecular Formula: C6H6N2S
InChi Key: XSRRCOBFMZWKJR-UHFFFAOYSA-N
SMILES: CC1=CSC(CC#N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results