accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1979-29-9

OOOFFF

CAS RN 1979-29-9

IUPAC Name: 2-(trifluoromethoxy)benzoate
Synonyms: 2-(trifluoromethoxy)benzoate
Molecular Weight (g/mol): 205.11
Molecular Formula: C8H4F3O3
InChi Key: JMYSPFGUBNENSE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC=C1OC(F)(F)F
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)