accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 197960-31-9

IUPAC Name: 2-cyano-5-methylbenzene-1-sulfonyl chloride
Synonyms: 2-cyano-5-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 215.65
Molecular Formula: C8H6ClNO2S
InChi Key: ZFQDREVAYVNITB-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C=C1)C#N)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results