accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 198-55-0

IUPAC Name: perylene
Synonyms: perylene dibenz(de,kl)anthracene peri-dinaphthalene α-perylene perilene peri dinaphthalene
Molecular Weight (g/mol): 252.32
Molecular Formula: C20H12
InChi Key: CSHWQDPOILHKBI-UHFFFAOYSA-N
SMILES: C1=CC2=C3C(C=CC=C3C3=C4C(C=CC=C24)=CC=C3)=C1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (8)
  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)