accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHN

CAS RN 19812-93-2

IUPAC Name: 4'-hydroxy-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-hydroxy-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: ZRMIETZFPZGBEB-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results