accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19814-75-6

IUPAC Name: 9,9-dimethyl-9H-xanthene
Synonyms: 9,9-dimethylxanthene
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: MTVNAPYHLASOSX-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2OC2=CC=CC=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results