accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19815-97-5

IUPAC Name: 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-5-yl)ethan-1-one
Synonyms: 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-5-yl)ethanone
Molecular Weight (g/mol): 257.08
Molecular Formula: C10H9BrO3
InChi Key: BSROYFIAEPSLCT-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=C2OCCOC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results