CAS RN 19819-95-5
IUPAC Name:
2-(2-chlorophenyl)ethan-1-ol
Synonyms:
2-(2-chlorophenyl)ethanol
Molecular Weight (g/mol):
156.61
Molecular Formula:
C8H9ClO
InChi Key:
IWNHTCBFRSCBQK-UHFFFAOYSA-N
SMILES:
OCCC1=CC=CC=C1Cl
1
–
1
of
1
results
1
2-Chlorophenethylalcohol, 98%
CAS: 19819-95-5 Molecular Formula: C8H9ClO Molecular Weight (g/mol): 156.61 MDL Number: MFCD00002888 InChI Key: IWNHTCBFRSCBQK-UHFFFAOYSA-N Synonym: 2-2-chlorophenyl ethanol,2-chlorophenethyl alcohol,benzeneethanol, 2-chloro,2-chlorophenethylalcohol,2-2-chlorophenyl ethan-1-ol,o-chlorophenethyl alcohol,o-chlorophenethylic alcohol,o-chlorophenylmethylcarbinol,chlorobenzene-ethanol,2-chlorobenzeneethanol PubChem CID: 88266 IUPAC Name: 2-(2-chlorophenyl)ethanol SMILES: C1=CC=C(C(=C1)CCO)Cl