accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1984-15-2

IUPAC Name: disodium hydrogen (hydrogen phosphonatomethyl)phosphonate
Synonyms: medronic acid
Molecular Weight (g/mol): 219.96
Molecular Formula: CH4Na2O6P2
InChi Key: RXSYPQBNTPQOTQ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OP([O-])(=O)CP(O)([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)