accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 198646-60-5

CH3H3CCH3OONOOHO

CAS RN 198646-60-5

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4-oxopiperidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
Molecular Weight (g/mol): 243.26
Molecular Formula: C11H17NO5
InChi Key: GPBCBXYUAJQMQM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(=O)CC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results