accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19875-60-6

CH3NCH3ONH(S)NCH3(R)HNOHOOHO(Z)

CAS RN 19875-60-6

IUPAC Name: (2Z)-but-2-enedioic acid; 3,3-diethyl-1-[(4S,7R)-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),2,9,12,14-pentaen-4-yl]urea
Synonyms: lisuride; maleic acid
Molecular Weight (g/mol): 454.53
Molecular Formula: C24H30N4O5
InChi Key: CVQFAMQDTWVJSV-BAXNFHPCSA-N
SMILES: OC(=O)\C=C/C(O)=O.CCN(CC)C(=O)N[C@@H]1CN(C)[C@@H]2CC3=CNC4=CC=CC(=C34)C2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results