CAS RN 1988-10-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy Propofol, TRC
CAS: 1988-10-9 Molecular Formula: C12 H18 O2 Molecular Weight (g/mol): 194.27 IUPAC Name: 2,6-di(propan-2-yl)benzene-1,4-diol SMILES: CC(C)c1cc(O)cc(C(C)C)c1O