accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 198904-31-3

CH3OONH(S)OHN(S)(S)OHNNNHO(S)HNOOCH3H3CCH3CH3H3CCH3H3C

CAS RN 198904-31-3

IUPAC Name: methyl N-[(1S)-1-{N'-[(2S,3S)-2-hydroxy-3-[(2S)-2-[(methoxycarbonyl)amino]-3,3-dimethylbutanamido]-4-phenylbutyl]-N'-{[4-(pyridin-2-yl)phenyl]methyl}hydrazinecarbonyl}-2,2-dimethylpropyl]carbamate
Synonyms: atazanavir ATV ATZ latazanavir atazanavirum zrivada
Molecular Weight (g/mol): 704.87
Molecular Formula: C38H52N6O7
InChi Key: AXRYRYVKAWYZBR-GASGPIRDSA-N
SMILES: COC(=O)N[C@H](C(=O)N[C@@H](CC1=CC=CC=C1)[C@@H](O)CN(CC1=CC=C(C=C1)C1=CC=CC=N1)NC(=O)[C@@H](NC(=O)OC)C(C)(C)C)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)