accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NSNH2NCl

CAS RN 19922-07-7

IUPAC Name: 3-(4-chlorophenyl)-1,2,4-thiadiazol-5-amine
Synonyms: 3-(4-chlorophenyl)-1,2,4-thiadiazol-5-amine
Molecular Weight (g/mol): 211.67
Molecular Formula: C8H6ClN3S
InChi Key: IUSHKZWNRIGLGO-UHFFFAOYSA-N
SMILES: NC1=NC(=NS1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results