accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ON

CAS RN 19924-43-7

IUPAC Name: 2-(3-methoxyphenyl)acetonitrile
Synonyms: 2-(3-methoxyphenyl)acetonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: LXKNAUOWEJWGTE-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CC#N)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)