accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHNONCH3H3CCl

CAS RN 19937-59-8

IUPAC Name: 1-(3-chloro-4-methoxyphenyl)-3,3-dimethylurea
Synonyms: metoxuron
Molecular Weight (g/mol): 228.68
Molecular Formula: C10H13ClN2O2
InChi Key: DSRNRYQBBJQVCW-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=O)N(C)C)C=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results