accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHNOHNNHN

CAS RN 19962-37-9

IUPAC Name: N-(6-oxo-6,7-dihydro-3H-purin-2-yl)acetamide
Synonyms: N-(6-oxo-3,7-dihydropurin-2-yl)acetamide
Molecular Weight (g/mol): 193.17
Molecular Formula: C7H7N5O2
InChi Key: MXSMRDDXWJSGMC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC(=O)C2=C(N1)N=CN2