accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHHO

CAS RN 20016-85-7

IUPAC Name: 2-hydroxybutanoic acid
Synonyms: α-hydroxybutyric acid (+/-)α-hydoxy butyric acid
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: AFENDNXGAFYKQO-UHFFFAOYNA-N
SMILES: CCC(O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)