accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2002-35-9

IUPAC Name: (2R,3S,5R)-2-(hydroxymethyl)-5-[6-(methylamino)-9H-purin-9-yl]oxolan-3-ol
Synonyms: N6-methyl-2'-deoxyadenosine N(6)-me-dado
Molecular Weight (g/mol): 265.27
Molecular Formula: C11H15N5O3
InChi Key: DYSDOYRQWBDGQQ-XLPZGREQSA-N
SMILES: CNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results