accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClONH2

CAS RN 20028-53-9

IUPAC Name: 2-amino-5-chlorobenzaldehyde
Synonyms: 2-amino-5-chlorobenzaldehyde
Molecular Weight (g/mol): 155.58
Molecular Formula: C7H6ClNO
InChi Key: KUIFMGITZFDQMP-UHFFFAOYSA-N
SMILES: NC1=C(C=O)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results