CAS RN 20041-90-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Caffeidine, TRC
CAS: 20041-90-1 Molecular Formula: C7 H12 N4 O Molecular Weight (g/mol): 168.2 Synonym: N,1-Dimethyl-4-(methylamino)imidazole-5-carboxamide,Imidazole-5-carboxamide, N,1-dimethyl-4-(methylamino)- (8CI),1H-Imidazole-5-carboxamide, N,1-dimethyl-4-(methylamino)- IUPAC Name: N,3-dimethyl-5-(methylamino)imidazole-4-carboxamide SMILES: CNC(=O)c1c(NC)ncn1C