accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 200422-18-0

(S)(R)(S)NO(R)(R)OOOOHHOHOHO

CAS RN 200422-18-0

IUPAC Name: 2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol hydrate
Synonyms: 2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol hydrate
Molecular Weight (g/mol): 319.27
Molecular Formula: C12H17NO9
InChi Key: KSEGJDPERYLUSV-UHFFFAOYNA-N
SMILES: O.OCC1OC(OC2=CC=C(C=C2)[N+]([O-])=O)C(O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results