accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20074-76-4

IUPAC Name: copper(2+) bis(prop-2-enoate)
Synonyms: copper(2+) diacrylate
Molecular Weight (g/mol): 205.66
Molecular Formula: C6H6CuO4
InChi Key: XPLSDXJBKRIVFZ-UHFFFAOYSA-L
SMILES: [Cu++].[O-]C(=O)C=C.[O-]C(=O)C=C
Percent Purity 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results