accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOClCl

CAS RN 2008-58-4

IUPAC Name: 2,6-dichlorobenzamide
Synonyms: 2,6-dichlorobenzamide BAM
Molecular Weight (g/mol): 190.02
Molecular Formula: C7H5Cl2NO
InChi Key: JHSPCUHPSIUQRB-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)