accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClClN

CAS RN 2008-75-5

IUPAC Name: hydrogen 1-(2-chloroethyl)piperidine chloride
Synonyms: hydrogen 1-(2-chloroethyl)piperidine chloride
Molecular Weight (g/mol): 184.1
Molecular Formula: C7H15Cl2N
InChi Key: VFLQQZCRHPIGJU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCCN1CCCCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)