CAS RN 2010-06-2
IUPAC Name:
4-phenyl-1,3-thiazol-2-amine
Synonyms:
phenthiazamine
Molecular Weight (g/mol):
176.24
Molecular Formula:
C9H8N2S
InChi Key:
PYSJLPAOBIGQPK-UHFFFAOYSA-N
SMILES:
NC1=NC(=CS1)C1=CC=CC=C1
1
–
2
of
2
results
1
2-Amino-4-phenylthiazole, 98%
CAS: 2010-06-2 Molecular Formula: C9H8N2S Molecular Weight (g/mol): 176.24 MDL Number: MFCD00039680 InChI Key: PYSJLPAOBIGQPK-UHFFFAOYSA-N Synonym: 4-phenylthiazol-2-amine,2-amino-4-phenylthiazole,phenthiazamine,2-thiazolamine, 4-phenyl,4-phenyl-2-thiazolamine,thiazole, 2-amino-4-phenyl,4-phenyl-thiazol-2-ylamine,4-phenyl-2-thiazolylamine,2-amino-4-phenyl-1,3-thiazole,4-phenyl-2-aminothiazole PubChem CID: 40302 IUPAC Name: 4-phenyl-1,3-thiazol-2-amine SMILES: C1=CC=C(C=C1)C2=CSC(=N2)N
2
2-Amino-4-phenylthiazole, 97%
CAS: 2010-06-2 Molecular Formula: C9H8N2S Molecular Weight (g/mol): 176.24 MDL Number: MFCD00039680 InChI Key: PYSJLPAOBIGQPK-UHFFFAOYSA-N