CAS RN 2013-12-9
CAS RN 2013-12-9
IUPAC Name:
(2R)-2-aminopentanoic acid
Synonyms:
D-(-)-norvaline D-ape D-nva
Molecular Weight (g/mol):
117.15
Molecular Formula:
C5H11NO2
InChi Key:
SNDPXSYFESPGGJ-SCSAIBSYSA-N
SMILES:
CCC[C@@H](N)C(O)=O
1
–
2
of
2
results
1
Thermo Scientific Chemicals D-Norvaline, 99%
CAS: 2013-12-9 Molecular Formula: C5H11NO2 Molecular Weight (g/mol): 117.15 MDL Number: MFCD00008097 InChI Key: SNDPXSYFESPGGJ-SCSAIBSYSA-N Synonym: d-norvaline,d--norvaline,2r-2-aminopentanoic acid,h-d-nva-oh,d---norvaline,d-2-aminovaleric acid,d-2-aminopentanoic acid,r-2-aminovaleric acid,r-2-aminopentanoic acid,d-ape PubChem CID: 439575 ChEBI: CHEBI:28804 IUPAC Name: (2R)-2-aminopentanoic acid SMILES: CCC[C@@H](N)C(O)=O
2
D-Norvaline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.