accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNN

CAS RN 20173-04-0

IUPAC Name: methyl[(pyridin-3-yl)methyl]amine
Synonyms: N-methyl-3-pyridylmethylamine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: MCSAQVGDZLPTBS-UHFFFAOYSA-N
SMILES: CNCC1=CC=CN=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)