CAS RN 20173-24-4
IUPAC Name:
2-(pyridin-3-yl)ethan-1-aminium
Synonyms:
2-(pyridin-3-yl)ethanaminium
Molecular Weight (g/mol):
123.18
Molecular Formula:
C7H11N2
InChi Key:
NAHHNSMHYCLMON-UHFFFAOYSA-O
SMILES:
[NH3+]CCC1=CC=CN=C1
1
–
1
of
1
results
1
3-(2-Aminoethyl)pyridine, 98%
CAS: 20173-24-4 Molecular Formula: C7H11N2 Molecular Weight (g/mol): 123.18 MDL Number: MFCD00191604 InChI Key: NAHHNSMHYCLMON-UHFFFAOYSA-O Synonym: 3-2-aminoethyl pyridine,2-pyridin-3-yl ethanamine,3-aminoethylpyridine,3-pyridineethanamine,2-pyridin-3-yl ethan-1-amine,2-3-pyridyl ethylamine,3-pyridineethaneamine,3-pyridine ethanamine,2-pyridin-3-yl-ethylamine,2-3-pyridinyl ethanamine PubChem CID: 854051 SMILES: [NH3+]CCC1=CC=CN=C1