CAS RN 2017555-09-6
CAS RN 2017555-09-6
IUPAC Name:
potassium trifluoro(4-{[(furan-2-yl)methyl]carbamoyl}phenyl)boranuide
Synonyms:
potassium trifluoro({4-[(furan-2-ylmethyl)carbamoyl]phenyl})boranuide
Molecular Weight (g/mol):
307.12
Molecular Formula:
C12H10BF3KNO2
InChi Key:
BNXMQAULUCBEPA-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CC=C(C=C1)C(=O)NCC1=CC=CO1
1
–
1
of
1
results
1
Potassium 4-(furfurylaminocarbonyl)phenyltrifluoroborate, Thermo Scientific™
CAS: 2017555-09-6 Molecular Formula: C12H10BF3KNO2 Molecular Weight (g/mol): 307.12 MDL Number: MFCD11977716 InChI Key: BNXMQAULUCBEPA-UHFFFAOYSA-N Synonym: potassium 4-furfurylamino-1-carbonyl phenyl trifluoroborate,potassium 4-furfurylaminocarbonyl phenyltrifluoroborate,amtb164,potassium 4-furfurylamino-1-carbonyl-phenyl trifluoroborate,potassium trifluoro 4-furan-2-yl methyl carbamoyl phenyl boranuide,potassium trifluoro 4-furan-2-ylmethyl carbamoyl phenyl boranuide PubChem CID: 71306500 IUPAC Name: potassium;trifluoro-[4-(furan-2-ylmethylcarbamoyl)phenyl]boranuide SMILES: [K+].F[B-](F)(F)C1=CC=C(C=C1)C(=O)NCC1=CC=CO1