CAS RN 201849-15-2
IUPAC Name:
2-bromo-1-chloro-4-fluorobenzene
Synonyms:
2-bromo-1-chloro-4-fluorobenzene
Molecular Weight (g/mol):
209.44
Molecular Formula:
C6H3BrClF
InChi Key:
FOCCSIJMXBTKHD-UHFFFAOYSA-N
SMILES:
FC1=CC(Br)=C(Cl)C=C1
1
–
1
of
1
results
1
2-Bromo-1-chloro-4-fluorobenzene, 98+%, Thermo Scientific™
CAS: 201849-15-2 Molecular Formula: C6H3BrClF Molecular Weight (g/mol): 209.44 MDL Number: MFCD00061120 InChI Key: FOCCSIJMXBTKHD-UHFFFAOYSA-N Synonym: 2-chloro-5-fluorobromobenzene,1-bromo-2-chloro-5-fluorobenzene,3-bromo-4-chlorofluorobenzene,2-bromo-1-chloro-4-fluoro-benzene,benzene, 2-bromo-1-chloro-4-fluoro,pubchem4270,acmc-209f6i,ksc494o8h,1-chloro-2-bromo-4-fluorobenzene,2-bromo-4-fluorochlorobenzene PubChem CID: 2773263 IUPAC Name: 2-bromo-1-chloro-4-fluorobenzene SMILES: FC1=CC(Br)=C(Cl)C=C1