accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2019997-40-9

IUPAC Name: 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furo[3,2-b]pyridine
Synonyms: 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furo[3,2-b]pyridine
Molecular Weight (g/mol): 245.09
Molecular Formula: C13H16BNO3
InChi Key: NCPRRWXGBQZDIU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=NC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results