CAS RN 20219-33-4
CAS RN 20219-33-4
IUPAC Name:
4-hydroxypent-3-en-2-one tetrakis(ethanol) tantalum
Synonyms:
4-hydroxypent-3-en-2-one tetrakis(ethyl alcohol) tantalum
Molecular Weight (g/mol):
465.34
Molecular Formula:
C13H32O6Ta
InChi Key:
YMZNJFSVFJDLIR-UHFFFAOYSA-N
SMILES:
[Ta].CCO.CCO.CCO.CCO.CC(O)=CC(C)=O
1
–
1
of
1
results
1
Tantalum(V) tetraethoxide 2,4-pentanedionate, 99.99%, (metals basis)
CAS: 20219-33-4 Molecular Formula: C13H27O6Ta Molecular Weight (g/mol): 460.30 MDL Number: MFCD00145349 InChI Key: LSALXRIVOAYYJW-GLUPTGNJSA-M Synonym: tantalum v tetraethoxide 2,4-pentanedionate PubChem CID: 122130355 SMILES: [Ta+5].CC[O-].CC[O-].CC[O-].CC[O-].C\C([O-])=C\C(C)=O