accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOH2NCl

CAS RN 202197-26-0

IUPAC Name: 3-chloro-4-[(3-fluorophenyl)methoxy]aniline
Synonyms: 3-chloro-4-[(3-fluorophenyl)methoxy]aniline
Molecular Weight (g/mol): 251.69
Molecular Formula: C13H11ClFNO
InChi Key: AYPFEYDGZDPAPE-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(OCC2=CC(F)=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results