accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 202522-22-3

NH3FFFF

CAS RN 202522-22-3

IUPAC Name: [4-fluoro-2-(trifluoromethyl)phenyl]methanaminium
Synonyms: [4-fluoro-2-(trifluoromethyl)phenyl]methanaminium
Molecular Weight (g/mol): 194.15
Molecular Formula: C8H8F4N
InChi Key: BLQGRYYLPWCHMA-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results