accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOO

CAS RN 20280-81-3

IUPAC Name: 4-methoxy-2H-chromen-2-one
Synonyms: 4-methoxychromen-2-one
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: MLCMXDYMSAZNPC-UHFFFAOYSA-N
SMILES: COC1=CC(=O)OC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results